Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 5A11]

Lieferant: OriGene
CF502666
ORIGCF502666EA 764 EUR
ORIGCF502666
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 5A11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1C2
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR