Anti-CLPP Mouse Monoclonal Antibody [clone: 1D3]

Lieferant: OriGene
CF502075
ORIGCF502075EA 764 EUR
ORIGCF502075
Anti-CLPP Mouse Monoclonal Antibody [clone: 1D3]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CLPP
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR