Anti-CD80 Mouse Monoclonal Antibody [clone: 3F1]

Lieferant: OriGene
CF501681
ORIGCF501681EA 764 EUR
ORIGCF501681
Anti-CD80 Mouse Monoclonal Antibody [clone: 3F1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR