Anti-PNMT Mouse Monoclonal Antibody [clone: 2H6]

Lieferant: OriGene
CF502824
ORIGCF502824EA 764 EUR
ORIGCF502824
Anti-PNMT Mouse Monoclonal Antibody [clone: 2H6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PNMT
Clonality: Monoclonal
Clone: 2H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR