Anti-PNMT Mouse Monoclonal Antibody [clone: 2A5]

Lieferant: OriGene
CF502818
ORIGCF502818EA 764 EUR
ORIGCF502818
Anti-PNMT Mouse Monoclonal Antibody [clone: 2A5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: PNMT
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR