Anti-SIL1 Mouse Monoclonal Antibody [clone: 1F9]

Lieferant: OriGene
CF501011
ORIGCF501011EA 764 EUR
ORIGCF501011
Anti-SIL1 Mouse Monoclonal Antibody [clone: 1F9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: SIL1
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR