Anti-LIPG Mouse Monoclonal Antibody [clone: 1B8]

Lieferant: OriGene
CF501002
ORIGCF501002EA 764 EUR
ORIGCF501002
Anti-LIPG Mouse Monoclonal Antibody [clone: 1B8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: LIPG
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR