Anti-MCL1 Mouse Monoclonal Antibody [clone: 3A12]

Lieferant: OriGene
CF500983
ORIGCF500983EA 764 EUR
ORIGCF500983
Anti-MCL1 Mouse Monoclonal Antibody [clone: 3A12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR