Anti-GBA3 Mouse Monoclonal Antibody [clone: 1F1]

Lieferant: OriGene
CF502602
ORIGCF502602EA 764 EUR
ORIGCF502602
Anti-GBA3 Mouse Monoclonal Antibody [clone: 1F1]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GBA3
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR