Anti-GSTT2 Mouse Monoclonal Antibody [clone: 5A11]

Lieferant: OriGene
CF501933
ORIGCF501933EA 786 EUR
ORIGCF501933
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 5A11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR