Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 1F11]

Lieferant: OriGene
CF501930
ORIGCF501930EA 764 EUR
ORIGCF501930
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 1F11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: EPM2AIP1
Clonality: Monoclonal
Clone: 1F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR