Anti-TRIM2 Mouse Monoclonal Antibody [clone: 2B2]

Lieferant: OriGene
ORIGCF501552EA 764 EUR
ORIGCF501552
Anti-TRIM2 Mouse Monoclonal Antibody [clone: 2B2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM2
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR