Anti-IVD Mouse Monoclonal Antibody [clone: 5A11]

Lieferant: OriGene
CF501544
ORIGCF501544EA 786 EUR
ORIGCF501544
Anti-IVD Mouse Monoclonal Antibody [clone: 5A11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IVD
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR