Anti-SRR Mouse Monoclonal Antibody [clone: 7G6]

Lieferant: OriGene
ORIGCF500958EA 764 EUR
ORIGCF500958
Anti-SRR Mouse Monoclonal Antibody [clone: 7G6]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SRR
Clonality: Monoclonal
Clone: 7G6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR