Anti-PSMC3 Mouse Monoclonal Antibody [clone: 7D1]

Lieferant: OriGene
CF500930
ORIGCF500930EA 786 EUR
ORIGCF500930
Anti-PSMC3 Mouse Monoclonal Antibody [clone: 7D1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMC3
Clonality: Monoclonal
Clone: 7D1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR