Anti-MEF2C Mouse Monoclonal Antibody [clone: 4B10]

Lieferant: OriGene
ORIGCF502566EA 764 EUR
ORIGCF502566
Anti-MEF2C Mouse Monoclonal Antibody [clone: 4B10]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MEF2C
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR