Anti-BIN3 Mouse Monoclonal Antibody [clone: 4A6]

Lieferant: OriGene
CF501913
ORIGCF501913EA 786 EUR
ORIGCF501913
Anti-BIN3 Mouse Monoclonal Antibody [clone: 4A6]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: BIN3
Clonality: Monoclonal
Clone: 4A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR