Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 7B5]

Lieferant: OriGene
CF501529
ORIGCF501529EA 764 EUR
ORIGCF501529
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 7B5]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BTN1A1
Clonality: Monoclonal
Clone: 7B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR