Anti-CAMK1D Mouse Monoclonal Antibody [clone: 2C6]

Lieferant: OriGene
ORIGCF500919EA 764 EUR
ORIGCF500919
Anti-CAMK1D Mouse Monoclonal Antibody [clone: 2C6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CAMK1D
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR