Anti-KCTD14 Mouse Monoclonal Antibody [clone: 3A4]

Lieferant: OriGene
ORIGCF502529EA 786 EUR
ORIGCF502529
Anti-KCTD14 Mouse Monoclonal Antibody [clone: 3A4]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KCTD14
Clonality: Monoclonal
Clone: 3A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR