Anti-ZFAND2B Mouse Monoclonal Antibody [clone: 2G10]

Lieferant: OriGene
ORIGCF502514EA 786 EUR
ORIGCF502514
Anti-ZFAND2B Mouse Monoclonal Antibody [clone: 2G10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ZFAND2B
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR