Anti-PBLD Mouse Monoclonal Antibody [clone: 4A5]

Lieferant: OriGene
CF501840
ORIGCF501840EA 764 EUR
ORIGCF501840
Anti-PBLD Mouse Monoclonal Antibody [clone: 4A5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PBLD
Clonality: Monoclonal
Clone: 4A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR