Anti-ATP5B Mouse Monoclonal Antibody [clone: 6C10]

Lieferant: OriGene
CF500838
ORIGCF500838EA 764 EUR
ORIGCF500838
Anti-ATP5B Mouse Monoclonal Antibody [clone: 6C10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ATP5B
Clonality: Monoclonal
Clone: 6C10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR