Anti-HAGHL Mouse Monoclonal Antibody [clone: 1G8]

Lieferant: OriGene
CF502476
ORIGCF502476EA 786 EUR
ORIGCF502476
Anti-HAGHL Mouse Monoclonal Antibody [clone: 1G8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HAGHL
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR