Anti-CRYAB Mouse Monoclonal Antibody [clone: 2A1]

Lieferant: OriGene
CF500550
ORIGCF500550EA 764 EUR
ORIGCF500550
Anti-CRYAB Mouse Monoclonal Antibody [clone: 2A1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR