Anti-BUB1B Mouse Monoclonal Antibody [clone: 11D7]

Lieferant: OriGene
CF500541
ORIGCF500541EA 764 EUR
ORIGCF500541
Anti-BUB1B Mouse Monoclonal Antibody [clone: 11D7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: BUB1B
Clonality: Monoclonal
Clone: 11D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR