Anti-IRF3 Mouse Monoclonal Antibody [clone: 2G3]

Lieferant: OriGene
CF500520
ORIGCF500520EA 786 EUR
ORIGCF500520
Anti-IRF3 Mouse Monoclonal Antibody [clone: 2G3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: IRF3
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR