Anti-HADH Mouse Monoclonal Antibody [clone: 1E6]

Lieferant: OriGene
TA802889
ORIGTA802889EA 616 EUR
ORIGTA802889
Anti-HADH Mouse Monoclonal Antibody [clone: 1E6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: HADH
Clonality: Monoclonal
Clone: 1E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR