Anti-CD5 Mouse Monoclonal Antibody [clone: 4F9]

Lieferant: OriGene
TA802411
ORIGTA802411EA 616 EUR
ORIGTA802411
Anti-CD5 Mouse Monoclonal Antibody [clone: 4F9]
Antikörper
Recommended Dilutions: Western Blot: 1:0

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR