Anti-NAB1 Mouse Monoclonal Antibody [clone: 2H3]

Lieferant: OriGene
ORIGTA803022EA 616 EUR
ORIGTA803022
Anti-NAB1 Mouse Monoclonal Antibody [clone: 2H3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NAB1
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR