Anti-CDCA7L Mouse Monoclonal Antibody [clone: 6C6]

Lieferant: OriGene
ORIGTA802969EA 616 EUR
ORIGTA802969
Anti-CDCA7L Mouse Monoclonal Antibody [clone: 6C6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CDCA7L
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR