Anti-HP Mouse monoclonal Antibody (Biotin) [clone: 6H2]

Lieferant: OriGene
TA700402
ORIGTA700402EA 647 EUR
ORIGTA700402
Anti-HP Mouse monoclonal Antibody (Biotin) [clone: 6H2]
Antikörper
Type: Primary
Antigen: HP
Clonality: monoclonal
Clone: 6H2
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR