Anti-TP63 Mouse monoclonal Antibody (Biotin) [clone: 5G7]

Lieferant: OriGene
TA700388
ORIGTA700388EA 647 EUR
ORIGTA700388
Anti-TP63 Mouse monoclonal Antibody (Biotin) [clone: 5G7]
Antikörper
Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: TP63
Clonality: monoclonal
Clone: 5G7
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR