Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 2A7]

Lieferant: OriGene
TA506919
ORIGTA506919EA 593 EUR
ORIGTA506919
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 2A7]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MADCAM1
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR