Anti-Ppargc1a Rabbit Polyclonal Antibody

Lieferant: OriGene
TA319007
ORIGTA319007EA 590 EUR
ORIGTA319007
Anti-Ppargc1a Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Ppargc1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR