Anti-Tlr3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318910
ORIGTA318910EA 590 EUR
ORIGTA318910
Anti-Tlr3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Tlr3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR