Anti-Nrxn3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318909
ORIGTA318909EA 634 EUR
ORIGTA318909
Anti-Nrxn3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:200-1000

Type: Primary
Antigen: Nrxn3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR