Anti-IgG Antibody (Gold, 10nm)

Lieferant: Boster
BSBTGA1004EA 221 EUR
BSBTGA1004
Anti-IgG Antibody (Gold, 10nm)
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: 10nm colloidal Gold
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR