Anti-Protein A Antibody (Biotin)

Lieferant: Boster
BSBTBA1025-1EA 0 EUR
BSBTBA1025-1
Anti-Protein A Antibody (Biotin)
Antikörper
Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR