Anti-IRAKM Rabbit Antibody

Lieferant: BosterBio
PA2207-1
BSBTPA2207-1EA 550 EUR
BSBTPA2207-1
Anti-IRAKM Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Interleukin-1 receptor-associated kinase 3(IRAK3) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IRAKM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR