Hematopoietic progenitor cell expansion medium DXF
Lieferant: PROMOCELL
C-28021 C-39891 C-39890
C-28021EA 252 EUR
C-28021 C-39891 C-39890
Hematopoietic progenitor cell expansion medium DXF
Zellkulturmedien
PromoCell Hematopoietic Progenitor Cell (HPC) Expansion Medium DXF was designed for the maximum expansion of primitive HPCs in a fully defined, xeno-free culture environment. This medium is chemically defined and free of all animal derived components and substances of human origin, with human serum albumin being the only exception.
NOTE: The medium must be supplemented with additional cytokines.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.