Schläuche, PFA, Chemfluor®
Lieferant: Saint-Gobain
VERNB2003.00X1.501EA 1460 EUR
VERNB2003.00X1.501 VERNB2004.00X1.001 VERNB2009.00X1.501 VERNB2010.00X1.001 VERNB2008.00X1.001 VERNB2006.00X1.001 VERNB2002.00X1.001
Schläuche, PFA, Chemfluor®
PFA, translucent.
  • High level of purity
  • Excellent chemical resistance
  • Retains higher mechanical strenght at elevated temperature compared to PTFE
  • High resistance to stress cracking
Chemfluor® PFA tubing is widely used in the semi-conductor, laboratory, enviromental and pharmaceutical industries where ultrapure chemicals (including water) require precise quality control.
Temperature resistance: up to +260 °C
Hardness (Shore durometer A): 60
Density: 2,17