Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Columbia Biosciences
COBSAP-114EA 281 EUR
COBSAP-114
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR