Anti-ZONA OCCLUDENS 1 alpha-minus Guinea pig Polyclonal Antibody

Lieferant: United States Biological
USBIZ0750-11AEA 707 EUR
USBIZ0750-11A
Anti-ZONA OCCLUDENS 1 alpha-minus Guinea pig Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:50; Western Blot: 1:50; Immunochemistry: 1:50

Type: Primary
Antigen: ZONA OCCLUDENS 1 alpha-minus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR