Anti-ZNF143 Mouse Monoclonal Antibody [clone: 12K197]

Lieferant: US Biological
Z0739-82
USBIZ0739-82EA 722 EUR
USBIZ0739-82
Anti-ZNF143 Mouse Monoclonal Antibody [clone: 12K197]
Antikörper
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: ZNF143 (zinc finger protein 143)
Clonality: monoclonal
Clone: 12K197
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR