Anti-SLC39A11 Rabbit Polyclonal Antibody

Lieferant: US Biological
Z0510-11
USBIZ0510-11EA 754 EUR
USBIZ0510-11
Anti-SLC39A11 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SLC39A11 (solute carrier family 39, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR