Anti-SLC39A4 Goat Polyclonal Antibody

Lieferant: US Biological
Z0510-05B
USBIZ0510-05BEA 420 EUR
USBIZ0510-05B
Anti-SLC39A4 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: SLC39A4 (solute carrier family 39 (zinc transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR