Anti-ZDHHC8 Goat Polyclonal Antibody

Lieferant: US Biological
Z0390-50B
USBIZ0390-50BEA 551 EUR
USBIZ0390-50B
Anti-ZDHHC8 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: ZDHHC8 (zinc finger, DHHC-type containing 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR