Anti-ZAP70 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 0.T.155]

Lieferant: US Biological
Z0300-05A
USBIZ0300-05AEA 647 EUR
USBIZ0300-05A
Anti-ZAP70 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 0.T.155]
Antikörper
Recommended dilution: Immunochemistry: 5-20 ?g/ml

Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: monoclonal
Clone: 0.T.155
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR