Anti-ZAP70 Rabbit Monoclonal Antibody [clone: 8A43]

Lieferant: US Biological
Z0300-03H
USBIZ0300-03HEA 554 EUR
USBIZ0300-03H
Anti-ZAP70 Rabbit Monoclonal Antibody [clone: 8A43]
Antikörper
Recommended dilution: Flow Cytometry: 1:200; Western Blot: 1:2000; Immunoprecipitation: 1:50

Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: monoclonal
Clone: 8A43
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR